missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
$39.14 - $180.81
Chemical Identifiers
| CAS | 2455-24-5 |
|---|---|
| Molecular Formula | C9H14O3 |
| Molecular Weight (g/mol) | 170.208 |
| MDL Number | MFCD00014515 |
| InChI Key | LCXXNKZQVOXMEH-UHFFFAOYSA-N |
| Synonym | tetrahydrofurfuryl methacrylate, sartomer sr 203, methacrylic acid tetrahydrofurfuryl ester, 2-propenoic acid, 2-methyl-, tetrahydro-2-furanyl methyl ester, tetrahydrofurfurylmethacrylate, tetrahydrofuran-2-yl methyl methacrylate, methacrylic acid, tetrahydrofurfuryl ester, tetrahydrofuran-2-ylmethyl 2-methylprop-2-enoate, sartomersr203, ageflex thfma |
| PubChem CID | 17151 |
| IUPAC Name | oxolan-2-ylmethyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCC1CCCO1 |
Chemical Identifiers
| 2455-24-5 | |
| 170.208 | |
| LCXXNKZQVOXMEH-UHFFFAOYSA-N | |
| 17151 | |
| CC(=C)C(=O)OCC1CCCO1 |
| C9H14O3 | |
| MFCD00014515 | |
| tetrahydrofurfuryl methacrylate, sartomer sr 203, methacrylic acid tetrahydrofurfuryl ester, 2-propenoic acid, 2-methyl-, tetrahydro-2-furanyl methyl ester, tetrahydrofurfurylmethacrylate, tetrahydrofuran-2-yl methyl methacrylate, methacrylic acid, tetrahydrofurfuryl ester, tetrahydrofuran-2-ylmethyl 2-methylprop-2-enoate, sartomersr203, ageflex thfma | |
| oxolan-2-ylmethyl 2-methylprop-2-enoate |
Specifications
| 2455-24-5 | |
| 85°C | |
| Clear Liquid at 20°C | |
| MFCD00014515 | |
| LCXXNKZQVOXMEH-UHFFFAOYSA-N | |
| oxolan-2-ylmethyl 2-methylprop-2-enoate | |
| 17151 | |
| ≥98.0% (GC) |
| Yellow | |
| 25 mL | |
| C9H14O3 | |
| tetrahydrofurfuryl methacrylate, sartomer sr 203, methacrylic acid tetrahydrofurfuryl ester, 2-propenoic acid, 2-methyl-, tetrahydro-2-furanyl methyl ester, tetrahydrofurfurylmethacrylate, tetrahydrofuran-2-yl methyl methacrylate, methacrylic acid, tetrahydrofurfuryl ester, tetrahydrofuran-2-ylmethyl 2-methylprop-2-enoate, sartomersr203, ageflex thfma | |
| CC(=C)C(=O)OCC1CCCO1 | |
| 170.208 | |
| 170.21 | |
| Tetrahydrofurfuryl Methacrylate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (5)-0057
TSCA : Yes