missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| Physical Form | Clear Liquid at 20°C |
| Boiling Point | 85°C |
| Formula Weight | 170.21 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | Tetrahydrofurfuryl Methacrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (5)-0057 |
| TSCA | Yes |