missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Bromide 95.0+%, TCI America™

Catalog No. T180825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T180825G 25 g
T1808100G 100 g
T1808500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T180825G Supplier TCI America Supplier No. T180825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1192-30-9
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD00016894
InChI Key VOHILFSOWRNVJJ-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
PubChem CID 14501
IUPAC Name 2-(bromomethyl)oxolane
SMILES BrCC1CCCO1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 70°C
Formula Weight 165.03
Percent Purity ≥95.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Bromide
EINECSNumber (9)-0494
RTECSNumber LU6000000
TSCA Yes
Recommended Storage Refrigerator