missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Alcohol 98.0+%, TCI America™
$39.14 - $67.11
Chemical Identifiers
| CAS | 97-99-4 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.13 |
| MDL Number | MFCD00005372 |
| InChI Key | BSYVTEYKTMYBMK-UHFFFAOYNA-N |
| Synonym | tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro |
| PubChem CID | 7360 |
| IUPAC Name | (oxolan-2-yl)methanol |
| SMILES | OCC1CCCO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T010625ML
|
TCI America
T010625ML |
25 mL |
Each for $39.14
|
|
|||||
|
T0106500ML
|
TCI America
T0106500ML |
500 mL |
Each for $67.11
|
|
|||||
Chemical Identifiers
| 97-99-4 | |
| 102.13 | |
| BSYVTEYKTMYBMK-UHFFFAOYNA-N | |
| 7360 | |
| OCC1CCCO1 |
| C5H10O2 | |
| MFCD00005372 | |
| tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro | |
| (oxolan-2-yl)methanol |
Specifications
| -80°C | |
| Yellow | |
| 25 mL | |
| C5H10O2 | |
| tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro | |
| OCC1CCCO1 | |
| 102.13 | |
| 102.13 | |
| Tetrahydrofurfuryl Alcohol |
| 97-99-4 | |
| 176°C | |
| Clear Liquid at 20°C | |
| MFCD00005372 | |
| BSYVTEYKTMYBMK-UHFFFAOYNA-N | |
| (oxolan-2-yl)methanol | |
| 7360 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (5)-0056
RTECSNumber : LU2450000
TSCA : Yes