missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Alcohol 98.0+%, TCI America™

Catalog No. T0106500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0106500ML 500 mL
T010625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0106500ML Supplier TCI America Supplier No. T0106500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97-99-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00005372
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
PubChem CID 7360
IUPAC Name (oxolan-2-yl)methanol
SMILES OCC1CCCO1

Specifications

Melting Point -80°C
Color Yellow
Quantity 500 mL
Physical Form Clear Liquid at 20°C
Boiling Point 176°C
Formula Weight 102.13
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Alcohol
EINECSNumber (5)-0056
RTECSNumber LU2450000
TSCA Yes