missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl alcohol, 99+%

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

Supplier:  Thermo Scientific Chemicals 420440010

 View more versions of this product

Catalog No. AC420440010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrahydrofurfuryl alcohol
99+%
1.0543g/mL
1 kg
Glass bottle
1.4510 to 1.4530
1.0543
MFCD00005372
17, V,3, 115
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
BSYVTEYKTMYBMK-UHFFFAOYNA-N
oxolan-2-ylmethanol
7360
99+%
-80.0°C
97-99-4
178.0°C
Authentic
Liquid
1.6% max. 1,2-Pentanediol (GC)
C5H10O2
6.24 mPa.s (20°C)
15,9358
Solubility in water: soluble. Other solubilities: miscible with alcohol, ether, acetone, chloroform, and benzene
OCC1CCCO1
102.13
102.13

Chemical Identifiers

97-99-4
102.13
BSYVTEYKTMYBMK-UHFFFAOYNA-N
7360
OCC1CCCO1
C5H10O2
MFCD00005372
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
oxolan-2-ylmethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o

GHS Signal Word: Danger

EINECSNumber : 202-625-6

RTECSNumber : LU2450000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only