missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Tetrahydrofurfuryl alcohol, 98%

Catalog No. AAA17078AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17078AE 100 mL
AAA17078AP 500 mL
AAA170780F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17078AE Supplier Thermo Scientific Chemicals Supplier No. A17078AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-99-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00005372
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
PubChem CID 7360
SMILES OCC1CCCO1

Specifications

Melting Point <-80°C
Density 1.053
Quantity 100 mL
Refractive Index 1.452
Beilstein 102723
Merck Index 14,9213
Boiling Point 178°C
IUPAC Name (oxolan-2-yl)methanol
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material (+/-)-Tetrahydrofurfuryl alcohol
Show More Show Less
Hazard Category H227-H319-H335-H360Df
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
EINECSNumber 202-625-6
RTECSNumber LU2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only