Learn More
Tetrahydrofurfuryl alcohol, 98%
CAS: 97-99-4 | C5H10O2 | 102.13 g/mol
$228.86 - $364.84
Chemical Identifiers
| CAS | 97-99-4 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.13 |
| MDL Number | MFCD00005372 |
| InChI Key | BSYVTEYKTMYBMK-UHFFFAOYNA-N |
| Synonym | tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro |
| PubChem CID | 7360 |
| SMILES | OCC1CCCO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC138180010
|
Thermo Scientific Chemicals
138180010 |
1 L | Glass bottle |
Each for $228.86
Case of 4 Each for $0.00
|
|
||||
|
AC138180025
|
Thermo Scientific Chemicals
138180025 |
2.5 L | Glass bottle |
Each for $364.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97-99-4 | |
| 102.13 | |
| BSYVTEYKTMYBMK-UHFFFAOYNA-N | |
| 7360 |
| C5H10O2 | |
| MFCD00005372 | |
| tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro | |
| OCC1CCCO1 |
Specifications
| -80.0°C | |
| 1.0543g/mL | |
| 1 L | |
| Glass bottle | |
| 1.4510 to 1.4530 | |
| 1.0543 | |
| MFCD00005372 | |
| 17, V,3, 115 | |
| tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro | |
| BSYVTEYKTMYBMK-UHFFFAOYNA-N | |
| (oxolan-2-yl)methanol | |
| 7360 | |
| 98% |
| 97-99-4 | |
| 178.0°C | |
| Authentic | |
| Liquid | |
| 97.5% min. (GC) | |
| C5H10O2 | |
| 6.24 mPa.s (20°C) | |
| 15, 9358 | |
| Solubility in water: soluble. Other solubilities: miscible with alcohol, ether, acetone, chloroform, and benzene | |
| OCC1CCCO1 | |
| 102.13 | |
| 102.13 | |
| Tetrahydrofurfuryl alcohol, 98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
GHS Signal Word: Danger
EINECSNumber : 202-625-6
RTECSNumber : LU2450000
TSCA : TSCA
RUO – Research Use Only