missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl acrylate, tech. 90%, stab.

Catalog No. aal1029918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1029918 50 g
AAL1029930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1029918 Supplier Thermo Scientific Chemicals Supplier No. L1029918
Only null left

CAS: 2399-48-6 | C8H12O3 | 156.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2399-48-6
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00014516
InChI Key YNXCGLKMOXLBOD-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester
PubChem CID 94232
IUPAC Name oxolan-2-ylmethyl prop-2-enoate
SMILES C=CC(=O)OCC1CCCO1

Specifications

Density 1.03
Boiling Point 88°C to 89°C (9 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.455
Quantity 50 g
UN Number UN3334
Beilstein 113115
Formula Weight 156.18
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetrahydrofurfuryl acrylate, stabilized with 500ppm hydroquinone
Show More Show Less
Hazard Category H227-H302-H314-H317-H360Df
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 219-268-7
RTECSNumber UD3670893
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only