missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. T223925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T223925G 25 g
T2239500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T223925G Supplier TCI America Supplier No. T223925G

Chemical Identifiers

CAS 2399-48-6
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
InChI Key YNXCGLKMOXLBOD-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester
PubChem CID 94232
IUPAC Name oxolan-2-ylmethyl prop-2-enoate
SMILES C=CC(=O)OCC1CCCO1

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 g
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydrofurfuryl Acrylate (stabilized with MEHQ)
EINECSNumber (5)-0057
RTECSNumber UD3670893
TSCA Yes