missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Acetate 97.0+%, TCI America™
$132.35 - $1058.78
Chemical Identifiers
| CAS | 637-64-9 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00022478 |
| InChI Key | AAQDYYFAFXGBFZ-UHFFFAOYSA-N |
| Synonym | tetrahydrofurfuryl acetate, 2-acetoxymethyloxolane, tetrahydrofuran-2-yl methyl acetate, 2-furanmethanol, tetrahydro-, acetate, tetrahydro-2-furanmethyl acetate, furfuryl alcohol, tetrahydro-, acetate, 2-acetoxymethyl tetrahydrofuran, tetrahydro-2-furanmethanol acetate, fema no. 3055, 2-furanmethanol, tetrahydro-, 2-acetate |
| PubChem CID | 12506 |
| IUPAC Name | oxolan-2-ylmethyl acetate |
| SMILES | CC(=O)OCC1CCCO1 |
Chemical Identifiers
| 637-64-9 | |
| 144.17 | |
| AAQDYYFAFXGBFZ-UHFFFAOYSA-N | |
| 12506 | |
| CC(=O)OCC1CCCO1 |
| C7H12O3 | |
| MFCD00022478 | |
| tetrahydrofurfuryl acetate, 2-acetoxymethyloxolane, tetrahydrofuran-2-yl methyl acetate, 2-furanmethanol, tetrahydro-, acetate, tetrahydro-2-furanmethyl acetate, furfuryl alcohol, tetrahydro-, acetate, 2-acetoxymethyl tetrahydrofuran, tetrahydro-2-furanmethanol acetate, fema no. 3055, 2-furanmethanol, tetrahydro-, 2-acetate | |
| oxolan-2-ylmethyl acetate |
Specifications
| 637-64-9 | |
| 195°C | |
| Clear Liquid at 20°C | |
| MFCD00022478 | |
| AAQDYYFAFXGBFZ-UHFFFAOYSA-N | |
| oxolan-2-ylmethyl acetate | |
| 12506 | |
| ≥97.0% (GC) |
| Colorless | |
| 25 g | |
| C7H12O3 | |
| tetrahydrofurfuryl acetate, 2-acetoxymethyloxolane, tetrahydrofuran-2-yl methyl acetate, 2-furanmethanol, tetrahydro-, acetate, tetrahydro-2-furanmethyl acetate, furfuryl alcohol, tetrahydro-, acetate, 2-acetoxymethyl tetrahydrofuran, tetrahydro-2-furanmethanol acetate, fema no. 3055, 2-furanmethanol, tetrahydro-, 2-acetate | |
| CC(=O)OCC1CCCO1 | |
| 144.17 | |
| 144.17 | |
| Tetrahydrofurfuryl Acetate |
Safety and Handling
EINECSNumber : (5)-0061
TSCA : Yes