missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-3-one 98.0+%, TCI America™

Catalog No. T29715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29715G 5 g
T29711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29715G Supplier TCI America Supplier No. T29715G

Chemical Identifiers

CAS 22929-52-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD07778393
InChI Key JLPJFSCQKHRSQR-UHFFFAOYSA-N
Synonym 3-oxotetrahydrofuran, dihydrofuran-3 2h-one, dihydro-3 2h-furanone, dihydrofuran-3-one, dihydro-furan-3-one, tetrahydrofuran-3-one, 3 2h-furanone, dihydro, 3-oxo-1,4-epoxybutane, tetrahydrofuran-3-yloxy
PubChem CID 529392
IUPAC Name oxolan-3-one
SMILES C1COCC1=O

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 1224
Boiling Point 68°C
Formula Weight 86.09
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrahydrofuran-3-one