missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. T134825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T134825G 25 g
T1348500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T134825G Supplier TCI America Supplier No. T134825G

Chemical Identifiers

CAS 16874-33-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00022473
InChI Key UJJLJRQIPMGXEZ-UHFFFAOYSA-N
Synonym 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
PubChem CID 86079
IUPAC Name oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 3265
Boiling Point 130°C
Formula Weight 116.12
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Tetrahydrofuran-2-carboxylic Acid