missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-pyran-4-one 95.0+%, TCI America™

Catalog No. p-7136969
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18061G 1 g
T18065G 5 g
T180625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T18061G Supplier TCI America Supplier No. T18061G
Only null left

Chemical Identifiers

CAS 29943-42-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00006581
InChI Key JMJRYTGVHCAYCT-UHFFFAOYSA-N
Synonym tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
PubChem CID 121599
IUPAC Name oxan-4-one
SMILES C1COCCC1=O

Specifications

Color Yellow
Boiling Point 69°C
Quantity 1 g
UN Number 1224
Formula Weight 100.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydro-4H-pyran-4-one