missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-pyran-4-one, 99%

Catalog No. AC189440050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189440050 5 g Glass bottle
AC189440250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189440050 Supplier Thermo Scientific Chemicals Supplier No. 189440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29943-42-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29943-42-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
InChI Key JMJRYTGVHCAYCT-UHFFFAOYSA-N
Synonym tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
PubChem CID 121599
IUPAC Name oxan-4-one
SMILES C1COCCC1=O

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 166.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Quantity 5 g
Beilstein 17,I,131
Specific Gravity 1.08
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrahydro-4H-pyran-4-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 249-967-2

RUO – Research Use Only