missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-pyran-4-one, 98%

Catalog No. AAA1901003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1901003 1 g
AAA1901006 5 g
AAA1901014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1901003 Supplier Thermo Scientific Chemicals Supplier No. A1901003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29943-42-8 | C5H8O2 | 100.117 g/mol

It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting group, synthesis of symmetric tetra substituted methanes. The methyl enol ether is a useful protecting agent for alcohols, e.g. in nucleotide synthesis, with the advantage over 3,4-Dihydro-2H-pyran. It is also employed in a study of the enantioselective alpha-aminoxylation of ketones with nitrosobenzene and L-proline in an ionic liquid. It undergoes condensation reactions in the preparation of dipeptides and spiroimidazolones. It is also employed in wittig reactions for the synthesis of Penicillins and in a ring of vitamin D3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the preparation of 4-methoxytetrahydropyran-4-yl protecting group, synthesis of symmetric tetra substituted methanes. The methyl enol ether is a useful protecting agent for alcohols, e.g. in nucleotide synthesis, with the advantage over 3,4-Dihydro-2H-pyran. It is also employed in a study of the enantioselective alpha-aminoxylation of ketones with nitrosobenzene and L-proline in an ionic liquid. It undergoes condensation reactions in the preparation of dipeptides and spiroimidazolones. It is also employed in wittig reactions for the synthesis of Penicillins and in a ring of vitamin D3

Solubility
Very soluble in water.

Notes
Store away from strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 29943-42-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00006581
InChI Key JMJRYTGVHCAYCT-UHFFFAOYSA-N
Synonym tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
PubChem CID 121599
IUPAC Name oxan-4-one
SMILES C1COCCC1=O

Specifications

Density 1.084
Boiling Point 165°C to 167°C
Flash Point 62°C (144°F)
Refractive Index 1.453
Quantity 1 g
Beilstein 106463
Solubility Information Very soluble in water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material Tetrahydro-4H-pyran-4-one
Hazard Category H226
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 249-967-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only