missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™
$59.99 - $59.99
Chemical Identifiers
| CAS | 5337-03-1 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.14 |
| InChI Key | AVPKHOTUOHDTLW-UHFFFAOYSA-N |
| Synonym | tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid |
| PubChem CID | 219302 |
Chemical Identifiers
| 5337-03-1 | |
| 130.14 | |
| tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid |
| C6H10O3 | |
| AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
| 219302 |
Specifications
| 5337-03-1 | |
| C6H10O3 | |
| tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid | |
| 130.14 | |
| 130.14 | |
| Tetrahydro-2H-pyran-4-carboxylic acid |
| 97% | |
| 1 g | |
| AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
| 219302 | |
| 97% |
Safety and Handling
GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.