missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™

Catalog No. CC29901DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29901DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC29901DA Supplier Thermo Scientific Chemicals Supplier No. CC29901DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
PubChem CID 219302

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material Tetrahydro-2H-pyran-4-carboxylic acid
Hazard Statement GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.