Learn More
Tetrahydro-2-furoic acid, 98+%
CAS: 16874-33-2 | C5H8O3 | 116.12 g/mol
$102.58 - $102.58
Chemical Identifiers
| CAS | 16874-33-2 |
|---|---|
| Molecular Formula | C5H8O3 |
| Molecular Weight (g/mol) | 116.12 |
| MDL Number | MFCD00022473 |
| InChI Key | UJJLJRQIPMGXEZ-UHFFFAOYSA-N |
| Synonym | 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088 |
| PubChem CID | 86079 |
| IUPAC Name | oxolane-2-carboxylic acid |
| SMILES | C1CC(OC1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC263140250
|
Thermo Scientific Chemicals
263140250 |
25 g | Glass bottle |
Each for $102.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16874-33-2 | |
| 116.12 | |
| UJJLJRQIPMGXEZ-UHFFFAOYSA-N | |
| 86079 | |
| C1CC(OC1)C(=O)O |
| C5H8O3 | |
| MFCD00022473 | |
| 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088 | |
| oxolane-2-carboxylic acid |
Specifications
| 16874-33-2 | |
| 99.0 | |
| 1.1700g/mL | |
| 25 g | |
| Glass bottle | |
| 1.4590 to 1.4610 | |
| 1.17 | |
| MFCD00022473 | |
| Solubility in water: soluble | |
| C1CC(OC1)C(=O)O | |
| 116.12 | |
| 116.12 | |
| Tetrahydro-2-furoic acid, 99+% |
| Yellow to Brown | |
| 100.0 | |
| 128.0°C to 129.0°C (13.0 mmHg) | |
| Authentic | |
| Liquid | |
| 98% min. (GC) | |
| C5H8O3 | |
| 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088 | |
| UJJLJRQIPMGXEZ-UHFFFAOYSA-N | |
| oxolane-2-carboxylic acid | |
| 86079 | |
| 98+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
TSCA : TSCA
RUO – Research Use Only