missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Tetrahydro-2-furoic acid, 97+%

Catalog No. AAB2292714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2292714 25 g
AAB2292722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2292714 Supplier Thermo Scientific Chemicals Supplier No. B2292714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16874-33-2 | C5H8O3 | 116.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16874-33-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00022473
InChI Key UJJLJRQIPMGXEZ-UHFFFAOYSA-N
Synonym 2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
PubChem CID 86079
IUPAC Name oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Density 1.213
Quantity 25 g
Odor Pungent
Refractive Index 1.46
UN Number UN3265
Beilstein 109671
Boiling Point 244°C
Formula Weight 116.12
Percent Purity ≥97%
Chemical Name or Material (+/-)-Tetrahydro-2-furoic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only