Learn More
Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 95%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Boiling Point | 60°C to 64°C (8 mmHg) |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 128.17 |
| Percent Purity | 95% |
| Chemical Name or Material | Tetrahydro-2,2-dimethyl-4H-pyran-4-one |
Safety and Handling
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only