missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium Iodide 98.0+%, TCI America™

Catalog No. p-7136549
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10105G 5 g
T101025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10105G Supplier TCI America Supplier No. T10105G

Chemical Identifiers

CAS 2138-24-1
Molecular Formula C24H52IN
Molecular Weight (g/mol) 481.591
MDL Number MFCD00041981
InChI Key VRKHAMWCGMJAMI-UHFFFAOYSA-M
Synonym tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
PubChem CID 75056
IUPAC Name tetrahexylazanium;iodide
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 481.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahexylammonium Iodide
EINECSNumber (2)-0186
TSCA Yes