missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium hydrogensulfate, For ion pair chromatography (T), ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013929
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013929 5 g
111013928 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013929 Supplier Merck Emd Millipore Supplier No. 872995GF

missing translation for 'validMainframeMsg'

Tetrahexylammonium hydrogensulfate is a quaternary ammonium salt mainly used as an electrolyte.

Chemical Identifiers

MDL Number MFCD00037675

Specifications

Melting Point 98°C to 100°C; 99°C to 102°C
CAS 32503-34-7
Linear Formula [CH3(CH2)5]4 N(HSO4)
Quantity 5 g
Formula Weight 451.75
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography