missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium Hydrogen Sulfate 98.0+%, TCI America™

Catalog No. p-7137770
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T34955G 5 g
T349525G 25 g
2 options

Catalog No. T34955G

Supplier: TCI America T34955G

Only null left

Chemical Identifiers

CAS 32503-34-7
Molecular Formula C24H52NO4S
Molecular Weight (g/mol) 450.74
MDL Number MFCD00037675
InChI Key RULHPTADXJPDSN-UHFFFAOYSA-L
Synonym tetrahexylammonium hydrogensulfate, tetrahexylammonium hydrogen sulfate, tetra-n-hexylammonium hydrogen sulfate, tetrahexylammonium hydrogensulphate, 1-hexanaminium, n,n,n-trihexyl-, sulfate 1:1, tetrahexylammonium hydrogen sulphate, tetrahexylazanium sulfate, acmc-1ailg, tetra-n-hexyl-ammonium hydrogensulphate, tetra-n-hexylammonium hydrogen sulphate
PubChem CID 11015848
IUPAC Name tetrahexylazanium sulfate
SMILES [O-]S([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 451.75
Percent Purity ≥98.0% (T,N)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahexylammonium Hydrogen Sulfate
EINECSNumber (2)-0186
TSCA Yes