Learn More
Tetrahexylammonium Bromide, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 4328-13-6 |
|---|---|
| Molecular Formula | C24H52BrN |
| Molecular Weight (g/mol) | 434.59 |
| MDL Number | MFCD00011858 |
| InChI Key | SYZCZDCAEVUSPM-UHFFFAOYSA-M |
| Synonym | tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1 |
| PubChem CID | 78026 |
| IUPAC Name | tetrahexylazanium;bromide |
| SMILES | [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC420400250
|
Thermo Scientific Chemicals
420400250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC420401000
|
Thermo Scientific Chemicals
420401000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystChemical Identifiers
| 4328-13-6 | |
| 434.59 | |
| SYZCZDCAEVUSPM-UHFFFAOYSA-M | |
| 78026 | |
| [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC |
| C24H52BrN | |
| MFCD00011858 | |
| tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1 | |
| tetrahexylazanium;bromide |
Specifications
| 4328-13-6 | |
| 98.5% min. (Argentometry) | |
| C24H52BrN | |
| MFCD00011858 | |
| tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1 | |
| [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC | |
| 434.59 | |
| 434.58 | |
| Tetrahexylammonium bromide |
| Authentic | |
| Glass bottle | |
| [CH3(CH2)5]4NBr | |
| 25g | |
| SYZCZDCAEVUSPM-UHFFFAOYSA-M | |
| tetrahexylazanium;bromide | |
| 78026 | |
| 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 224-363-1
RUO â Research Use Only