missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluoroboric acid, ca 50% w/w aq. soln.
Description
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
Specifications
Specifications
| Concentration | ≈50% w/w aq. soln. |
| Boiling Point | 130°C (decomposition) |
| Density | 1.4 |
| Odor | Pungent |
| Quantity | 500 g |
| Solubility Information | Miscible with water. |
| Formula Weight | 87.81 |
| Concentration or Composition (by Analyte or Components) | ≈50% w/w aq. soln. |
| Chemical Name or Material | Tetrafluoroboric acid |
Safety and Handling
| Hazard Category | H314-H360FD |
| Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 240-898-3 |
| RTECSNumber | ED2685000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only