missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroboric acid, ca 50% w/w aq. soln.

Catalog No. AAL1403722
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1403722 100 g
AAL1403736 500 g
AAL140370E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1403722 Supplier Thermo Scientific Chemicals Supplier No. L1403722

CAS: 16872-11-0

Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and acids.

Chemical Identifiers

CAS 16872-11-0
MDL Number MFCD00011345
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq

Specifications

Boiling Point 130°C (decomposition)
Concentration ≈50% w/w aq. soln.
Density 1.4
Odor Pungent
Quantity 100 g
Solubility Information Miscible with water.
Formula Weight 87.81
Concentration or Composition (by Analyte or Components) ≈50% w/w aq. soln.
Chemical Name or Material Tetrafluoroboric acid
Hazard Category H314-H360FD
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 240-898-3
RTECSNumber ED2685000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only