missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylgermanium, 99%

CAS: 597-63-7 | C8H20Ge | 188.878 g/mol

$64.44 - $64.44

Chemical Identifiers

CAS 597-63-7
Molecular Formula C8H20Ge
Molecular Weight (g/mol) 188.878
MDL Number MFCD00015096
InChI Key QQXSEZVCKAEYQJ-UHFFFAOYSA-N
Synonym tetraethylgermanium, germane, tetraethyl, tetraethyl germane, germanium, tetraethyl, acmc-209mfg, c2h5 4ge, qqxsezvckaeyqj-uhfffaoysa, wln: 2-ge-2&2&2
PubChem CID 11703
IUPAC Name tetraethylgermane
SMILES CC[Ge](CC)(CC)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2762503
View Documents
Thermo Scientific Chemicals
H2762503
1 g
Each for $64.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

597-63-7
188.878
QQXSEZVCKAEYQJ-UHFFFAOYSA-N
11703
CC[Ge](CC)(CC)CC
C8H20Ge
MFCD00015096
tetraethylgermanium, germane, tetraethyl, tetraethyl germane, germanium, tetraethyl, acmc-209mfg, c2h5 4ge, qqxsezvckaeyqj-uhfffaoysa, wln: 2-ge-2&2&2
tetraethylgermane

Specifications

597-63-7
163°C to 164°C
C8H20Ge
MFCD00015096
UN1993
QQXSEZVCKAEYQJ-UHFFFAOYSA-N
tetraethylgermane
11703
99%
0.998
35°C (95°F)
1.442
1 g
tetraethylgermanium, germane, tetraethyl, tetraethyl germane, germanium, tetraethyl, acmc-209mfg, c2h5 4ge, qqxsezvckaeyqj-uhfffaoysa, wln: 2-ge-2&2&2
CC[Ge](CC)(CC)CC
188.878
188.88
Tetraethylgermanium
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 209-905-7

RTECSNumber : LY5290000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only