missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyleneglycol monomethyl ether, 98%

Catalog No. p-3830333
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420350050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420350050 Supplier Thermo Scientific Chemicals Supplier No. 420350050
Only null left

CAS: 23783-42-8 | C9H20O5 | 208.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23783-42-8
Molecular Formula C9H20O5
Molecular Weight (g/mol) 208.25
MDL Number MFCD00041756
InChI Key ZNYRFEPBTVGZDN-UHFFFAOYSA-N
Synonym 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol
PubChem CID 90263
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCO

Specifications

Color Colorless to Yellow
Density 0.9870g/mL
Boiling Point 158°C to 160°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4440 to 1.446
Linear Formula CH3O(CH2CH2O)3CH2CH2OH
Quantity 5 g
Specific Gravity 0.987
Formula Weight 208.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetraethyleneglycol monomethyl ether, 98%
Show More Show Less
EINECSNumber 245-883-5
TSCA TSCA

RUO – Research Use Only