missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene Glycol Diacrylate (stabilized with MEHQ) 90.0+%, TCI America™

$54.06 - $167.76

Chemical Identifiers

CAS 17831-71-9
Molecular Formula C14H22O7
Molecular Weight (g/mol) 302.323
MDL Number MFCD00008630
InChI Key HCLJOFJIQIJXHS-UHFFFAOYSA-N
PubChem CID 28803
IUPAC Name 2-[2-[2-(2-prop-2-enoyloxyethoxy)ethoxy]ethoxy]ethyl prop-2-enoate
SMILES C=CC(=O)OCCOCCOCCOCCOC(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T156925G
View Documents
TCI America
T156925G
25 g
Each for $54.06
Only null left
 
T1569500G
View Documents
TCI America
T1569500G
500 g
Each for $167.76
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17831-71-9
302.323
HCLJOFJIQIJXHS-UHFFFAOYSA-N
2-[2-[2-(2-prop-2-enoyloxyethoxy)ethoxy]ethoxy]ethyl prop-2-enoate
C14H22O7
MFCD00008630
28803
C=CC(=O)OCCOCCOCCOCCOC(=O)C=C

Specifications

17831-71-9
C14H22O7
25 g
HCLJOFJIQIJXHS-UHFFFAOYSA-N
2-[2-[2-(2-prop-2-enoyloxyethoxy)ethoxy]ethoxy]ethyl prop-2-enoate
28803
≥90.0% (GC)
Tetraethylene Glycol Diacrylate (stabilized with MEHQ)
Yellow
MFCD00008630
1760
C=CC(=O)OCCOCCOCCOCCOC(=O)C=C
302.323
302.32
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0152

RTECSNumber : AS8100000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'