missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylene glycol, 99.5%

CAS: 112-60-7 | C8H18O5 | 194.23 g/mol
$124.78 - $124.78
Chemical Identifiers
CAS | 112-60-7 |
---|---|
Molecular Formula | C8H18O5 |
Molecular Weight (g/mol) | 194.23 |
InChI Key | UWHCKJMYHZGTIT-UHFFFAOYSA-N |
Synonym | tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843Show More |
PubChem CID | 8200 |
ChEBI | CHEBI:44920 |
IUPAC Name | 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol |
SMILES | C(COCCOCCOCCO)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC149590010
|
Thermo Scientific Chemicals
149590010 |
1 L | Glass Bottle |
Each for $124.78
Case of 4 Each for $499.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-60-7 | |
194.23 | |
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843Show More | |
CHEBI:44920 | |
C(COCCOCCOCCO)O |
C8H18O5 | |
UWHCKJMYHZGTIT-UHFFFAOYSA-N | |
8200 | |
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol |
Specifications
112-60-7 | |
Yellow | |
324.0°C to 330.0°C | |
Authentic | |
Glass Bottle | |
1.4580 to 1.4600 | |
1 L | |
1.12 | |
Solubility in water: soluble. Other solubilities: insoluble in benzene and toluene, practically insoluble in diethylether | |
C(COCCOCCOCCO)O | |
194.23 | |
44.9 mPa.s (25°C) | |
194.23 | |
Liquid |
-4.1°C | |
1.1200g/mL | |
176°C | |
99.4% min. (GC) | |
C8H18O5 | |
O(CH2CH2OCH2CH2OH)2 | |
01,468 | |
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843 | |
UWHCKJMYHZGTIT-UHFFFAOYSA-N | |
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol | |
8200 | |
CHEBI:44920 | |
99.5% | |
Tetraethylene glycol |
Safety and Handling
EINECSNumber : 203-989-9
RUO – Research Use Only