missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Trifluoromethanesulfonate 99.0+%, TCI America™

Catalog No. p-7136940
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17455G 5 g
T174525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17455G Supplier TCI America Supplier No. T17455G
Only null left

Chemical Identifiers

CAS 35895-69-3
Molecular Formula C9H20F3NO3S
Molecular Weight (g/mol) 279.318
MDL Number MFCD00042586
InChI Key PUZYNDBTWXJXKN-UHFFFAOYSA-M
Synonym Tetraethylammonium Triflate
PubChem CID 2776566
IUPAC Name tetraethylazanium;trifluoromethanesulfonate
SMILES CC[N+](CC)(CC)CC.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 279.32
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Trifluoromethanesulfonate