missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7136533
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09835G 5 g
T098325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09835G Supplier TCI America Supplier No. T09835G
Only null left

Chemical Identifiers

CAS 429-06-1
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00011827
InChI Key XJRAKUDXACGCHA-UHFFFAOYSA-N
Synonym tetraethylammonium tetrafluoroborate, tetraethylazanium tetrafluoroborate, tetraethylammoniumtetrafluoroborate, acmc-1ago3, ksc238k1t, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-, ethanaminium, n,n,n-triethyl-, tetrafluoroborate 1-1:1, tetraethyl ammonium tetrafluoroborate, tetraethylammonium tetra fluoro borate
PubChem CID 2724277
IUPAC Name tetraethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CC[N+](CC)(CC)CC

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 217.06
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Tetrafluoroborate
EINECSNumber (2)-0186
TSCA Yes