missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium perchlorate, 0.2M aqueous solution

Catalog No. AC379801000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379801000 100 mL Plastic bottle
AC379805000 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC379801000 Supplier Thermo Scientific Chemicals Supplier No. 379801000

CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst

Specifications

Chemical Name or Material Tetraethylammonium perchlorate
CAS 2567-83-1, 7732-18-5
Density 1.0016g/mL
Packaging Plastic bottle
Molecular Formula C8H20ClNO4
Quantity 100 mL
MDL Number MFCD00041977
Specific Gravity 1.0016
Synonym tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry
InChI Key WGHUNMFFLAMBJD-UHFFFAOYSA-M
SMILES [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;perchlorate
Molecular Weight (g/mol) 229.70
PubChem CID 2734932
Formula Weight 229.7
Show More Show Less

RUO – Research Use Only