missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide (35% in Water), TCI America™

Catalog No. T258925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T258925ML 25 mL
T2589500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T258925ML Supplier TCI America Supplier No. T258925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
IUPAC Name tetraethylazanium hydroxide
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Color Yellow
Quantity 25 mL
UN Number 3267
Formula Weight 147.26
Physical Form Liquid
Chemical Name or Material Tetraethylammonium Hydroxide (35% in Water)
EINECSNumber (2)-0186
RTECSNumber KH3150000
TSCA Yes