Learn More
Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade
CAS: 77-98-5 | C8H21NO | 147.26 g/mol
Supplier: Thermo Scientific Chemicals 04302322
Description
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetraethylammonium hydroxide | |
| 1.023 g/mL | |
| Amine-like | |
| 1.404 | |
| MFCD00009024 | |
| UN3267 | |
| Air Sensitive | |
| tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
| LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
| tetraethylazanium hydroxide | |
| 6509 | |
| Reagent | |
| Liquid |
| 77-98-5 | |
| (decomposition) | |
| C8H21NO | |
| (CH3CH2)4NOH | |
| 100 g | |
| 3912961 | |
| 14,9201 | |
| Miscible with water. | |
| [OH-].CC[N+](CC)(CC)CC | |
| 147.26 | |
| 147.26 | |
| 35% w/w aq. soln. |
Chemical Identifiers
| 77-98-5 | |
| 147.26 | |
| LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
| 6509 |
| C8H21NO | |
| MFCD00009024 | |
| tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
| [OH-].CC[N+](CC)(CC)CC |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H300-H311-H314-H335-H370-H372
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 201-073-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only