missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade

Catalog No. AA4302336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4302336 500 g
AA4302314 25 g
AA4302322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4302336 Supplier Thermo Scientific Chemicals Supplier No. 04302336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Density 1.023 g/mL
Boiling Point (decomposition)
Odor Amine-like
Refractive Index 1.404
Linear Formula (CH3CH2)4NOH
Quantity 500 g
UN Number UN3267
Beilstein 3912961
Sensitivity Air Sensitive
Merck Index 14,9201
Solubility Information Miscible with water.
IUPAC Name tetraethylazanium;hydroxide
Formula Weight 147.26
Grade Reagent
Concentration or Composition (by Analyte or Components) 35% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetraethylammonium hydroxide
Show More Show Less
Hazard Category H290-H300-H311-H314-H335-H370-H372
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 201-073-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only