missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 25% w/w in methanol

Catalog No. p-7049622
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1632818 50 g
AA1632830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1632818 Supplier Thermo Scientific Chemicals Supplier No. 01632818
Only null left

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

It is used as a phase transfer catalyst, the template agent synthesized by molecular sieve and cleaning agents, as a surfactant, used to anisotropically etch silicon, in thermochemolysis, as a basic solvent in the development of acidic photo resist in the photolithography process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-98-5
Molecular Formula C8H21NO
Molecular Weight (g/mol) 147.26
MDL Number MFCD00009024
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
PubChem CID 6509
IUPAC Name tetraethylazanium;hydroxide
SMILES [OH-].CC[N+](CC)(CC)CC

Specifications

Density 0.88 g/mL
Boiling Point (decomposition)
Flash Point 16°C (60°F)
Odor Like alcohol
Refractive Index 1.38
Quantity 50 g
UN Number UN2924
Beilstein 3912961
Sensitivity Air Sensitive
Merck Index 14,9201
Solubility Information Soluble in water.
Formula Weight 147.26
Concentration or Composition (by Analyte or Components) 25% w/w in methanol
Physical Form Liquid
Chemical Name or Material Tetraethylammonium hydroxide
Show More Show Less
Hazard Category H225-H290-H300-H311+H331-H314-H335-H370-H372
Hazard Statement GHS H Statement
H224-H300-H310-H330-H370-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 201-073-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only