Learn More
Tetraethylammonium fluoride hydrate, 97%
CAS: 98330-04-2 | C8H20FN | 149.25 g/mol
$156.07 - $1,906.53
Chemical Identifiers
| CAS | 98330-04-2 |
|---|---|
| Molecular Formula | C8H20FN |
| Molecular Weight (g/mol) | 149.25 |
| MDL Number | MFCD00149990 |
| InChI Key | QSUJAUYJBJRLKV-UHFFFAOYSA-M |
| Synonym | tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate |
| PubChem CID | 16211652 |
| IUPAC Name | tetraethylazanium;fluoride;hydrate |
| SMILES | [F-].CC[N+](CC)(CC)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2317306
|
Thermo Scientific Chemicals
B2317306 |
5 g |
N/A
|
|
|||||
|
AAB2317314
|
Thermo Scientific Chemicals
B2317314 |
25 g |
N/A
|
|
|||||
|
AAB2317322
|
Thermo Scientific Chemicals
B2317322 |
100 g |
N/A
|
|
|||||
Description
Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.
Solubility
Soluble in water.
Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 98330-04-2 | |
| 149.25 | |
| QSUJAUYJBJRLKV-UHFFFAOYSA-M | |
| 16211652 | |
| [F-].CC[N+](CC)(CC)CC |
| C8H20FN | |
| MFCD00149990 | |
| tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate | |
| tetraethylazanium;fluoride;hydrate |
Specifications
| 98330-04-2 | |
| MFCD00149990 | |
| 4152659 | |
| tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate | |
| QSUJAUYJBJRLKV-UHFFFAOYSA-M | |
| tetraethylazanium;fluoride;hydrate | |
| 16211652 | |
| 97% |
| C8H20FN | |
| 5 g | |
| Hygroscopic | |
| Soluble in water. | |
| [F-].CC[N+](CC)(CC)CC | |
| 149.25 | |
| 149.26 (Anhydrous) | |
| Tetraethylammonium fluoride hydrate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-559-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only