missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium fluoride hydrate, 97%

Catalog No. AAB2317306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2317306 5 g
AAB2317314 25 g
AAB2317322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2317306 Supplier Thermo Scientific Chemicals Supplier No. B2317306

CAS: 98330-04-2 | C8H20FN | 149.25 g/mol

Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium fluoride hydrate is used as a catalyst for oxidation, methylation and deuteration reactions of fluorine. It is involved as a reagent for the cleavage of Si-C bonds and also acts as a phase transfer catalyst in synthetic chemistry.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 98330-04-2
Molecular Formula C8H20FN
Molecular Weight (g/mol) 149.25
MDL Number MFCD00149990
InChI Key QSUJAUYJBJRLKV-UHFFFAOYSA-M
Synonym tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate
PubChem CID 16211652
IUPAC Name tetraethylazanium;fluoride;hydrate
SMILES [F-].CC[N+](CC)(CC)CC

Specifications

Quantity 5 g
Beilstein 4152659
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 149.26 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Tetraethylammonium fluoride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-559-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only