missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium Chloride 98.0+%, TCI America™
Supplier: TCI America T0095100G
Specifications
| Tetraethylammonium Chloride | |
| White | |
| C8H20ClN | |
| 100 g | |
| YMBCJWGVCUEGHA-UHFFFAOYSA-M | |
| tetraethylazanium chloride | |
| 5946 | |
| 165.71 | |
| Crystalline Powder |
| 56-34-8 | |
| 4.0 to 7.0 (50g/L H2O 25°C) | |
| MFCD00149992 | |
| tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn | |
| [Cl-].CC[N+](CC)(CC)CC | |
| 165.71 | |
| CHEBI:78161 | |
| ≥98.0% (T) |
Chemical Identifiers
| 56-34-8 | |
| 165.71 | |
| YMBCJWGVCUEGHA-UHFFFAOYSA-M | |
| 5946 | |
| tetraethylazanium chloride |
| C8H20ClN | |
| MFCD00149992 | |
| tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn | |
| CHEBI:78161 | |
| [Cl-].CC[N+](CC)(CC)CC |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS6125000
TSCA : Yes