missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Chloride 98.0+%, TCI America™

Catalog No. T0095500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0095500G 500 g
T009525G 25 g
T0095100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0095500G Supplier TCI America Supplier No. T0095500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-34-8
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.71
MDL Number MFCD00149992
InChI Key YMBCJWGVCUEGHA-UHFFFAOYSA-M
Synonym tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn
PubChem CID 5946
ChEBI CHEBI:78161
IUPAC Name tetraethylazanium chloride
SMILES [Cl-].CC[N+](CC)(CC)CC

Specifications

Color White
pH 4.0 to 7.0 (50g/L H2O 25°C)
Quantity 500 g
Formula Weight 165.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Chloride
EINECSNumber (2)-0186
RTECSNumber BS6125000
TSCA Yes