missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7136076
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
T009425G 25 g
T0094500G 500 g
2 options

Catalog No. T009425G

Supplier: TCI America T009425G

Only null left

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
MDL Number MFCD00011825
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 285°C
Color White
pH 4.0 to 7.0 (50g/L H2O 25°C)
Quantity 25 g
Formula Weight 210.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Bromide
EINECSNumber (2)-0186
RTECSNumber BS5950000
TSCA Yes