missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium bromide, 98%

CAS: 71-91-0 | C8H20BrN | 210.159 g/mol

Supplier:  Thermo Scientific Chemicals A138350B

 View more versions of this product

Catalog No. AAA138350B


Only null left
Explore available promotions

Description

Description

Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium bromide is used as a phase transfer catalyst. It acts as a source of tetraethylammonium ions in pharmacological and physiological studies. It is also employed in organic chemical synthesis. It plays a major role in the preparation of tetraethylammonium superoxide from potassium superoxide for the conversion of primary alkyl halides to dialkyl peroxides. Furthermore, It is utilized as a catalyst in the oxidation of organic sulfide to sulfoxide using o-iodoxybenzoic acid.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

Tetraethylammonium bromide
∼285°C (decomposition)
Mild
MFCD00011825
3563430
14,9199
Soluble in water.
CC[N+](CC)(CC)CC.[Br-]
210.159
210.16
71-91-0
1.397
C8H20BrN
1000 g
Hygroscopic
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
HWCKGOZZJDHMNC-UHFFFAOYSA-M
tetraethylazanium;bromide
6285
98%

Chemical Identifiers

71-91-0
210.159
HWCKGOZZJDHMNC-UHFFFAOYSA-M
6285
CC[N+](CC)(CC)CC.[Br-]
C8H20BrN
MFCD00011825
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
tetraethylazanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 200-769-4

RTECSNumber : BS5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only