missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium acetate tetrahydrate, 96%

Catalog No. AC194940250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC194940250 25 g Glass bottle
AC194941000 100 g Glass bottle
AC194945000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC194940250 Supplier Thermo Scientific Chemicals Supplier No. 194940250

Phase transfer catalyst | CAS: 67533-12-4 | C10H23NO2·4H2O | 261.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 67533-12-4
Molecular Formula C10H23NO2·4H2O
Molecular Weight (g/mol) 261.35
InChI Key QNOAKQGNSWGYOE-UHFFFAOYSA-M
Synonym tetraethylammonium acetate tetrahydrate, ethanaminium, n,n,n-triethyl-, acetate, tetrahydrate, tetraethylazanium acetate tetrahydrate, tetraethylazanium ethanoate tetrahydrate, acetate ion tetraethylammonium tetrahydrate, ethanaminium,n,n,n-triethyl-,acetate,tetrahydrate
PubChem CID 2724098
IUPAC Name tetraethylazanium;acetate;tetrahydrate
SMILES CC[N+](CC)(CC)CC.CC(=O)[O-].O.O.O.O

Specifications

Melting Point 40.0°C to 45.0°C
Color Colorless to White
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (C2H5)4N(O2CCH3)·4H2O
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 261.35
Percent Purity 97+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium acetate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only