missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl Orthosilicate 97.0+%, TCI America™

Catalog No. T010025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T010025ML 25 mL
T0100500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T010025ML Supplier TCI America Supplier No. T010025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-10-4
Molecular Formula C8H20O4Si
Molecular Weight (g/mol) 208.329
MDL Number MFCD00009062
InChI Key BOTDANWDWHJENH-UHFFFAOYSA-N
Synonym tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
PubChem CID 6517
IUPAC Name tetraethyl silicate
SMILES CCO[Si](OCC)(OCC)OCC

Specifications

Melting Point -82°C
Color Colorless
Boiling Point 165°C
Quantity 25 mL
UN Number 1292
Formula Weight 208.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethyl Orthosilicate
EINECSNumber (2)-2048
RTECSNumber VV9450000
TSCA Yes