missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl orthosilicate, 98%

CAS: 78-10-4 | C8H20O4Si | 208.33 g/mol

Supplier:  Thermo Scientific Chemicals 157810010

 View more versions of this product

Catalog No. AC157810010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetraethyl orthosilicate
-77.0°C
166.0°C to 169.0°C
Authentic
Glass bottle
1.3820 to 1.3840
MFCD00009062
01,334
15,3903
Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, hydrocarbons, aromatics,, esters, ketones,, slightly soluble in benzene
CCO[Si](OCC)(OCC)OCC
208.33
0.75 mPa.s (20°C)
98%
78-10-4
0.9400g/mL
45°C
1% max. Ethanol (GC)
C8H20O4Si
Si(OC2H5)4
1 L
0.94
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
BOTDANWDWHJENH-UHFFFAOYSA-N
tetraethyl silicate
6517
208.33
Liquid

Chemical Identifiers

78-10-4
208.33
BOTDANWDWHJENH-UHFFFAOYSA-N
6517
CCO[Si](OCC)(OCC)OCC
C8H20O4Si
MFCD00009062
tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
tetraethyl silicate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-083-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only