missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Tetraethyl orthosilicate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$482.45 - $482.45
Specifications
| CAS | 78-10-4 |
|---|---|
| Density | 0.933 g/mL (at 20°C) |
| Boiling Point | 168°C |
| Molecular Formula | C8H20O4Si |
| Linear Formula | Si(OC2H5)4 |
Description
Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.Specifications
| 78-10-4 | |
| 168°C | |
| Si(OC2H5)4 | |
| MFCD00009062 | |
| 208.33 | |
| Respiratory system |
| 0.933 g/mL (at 20°C) | |
| C8H20O4Si | |
| n20/D 1.382 (literature) | |
| Orthosilicic acid tetraethyl ester; Silicon tetraethoxide; TEOS; Tetraethoxysilane; Tetraethoxysilicon(IV); Tetraethyl silicate; Orthosilicic acid tetraethyl ester; TEOS; Tetraethoxysilane | |
| 98% |