missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Tetraethyl orthosilicate
Description
Will interact with dodecylamine in the formation of intercalation compounds of H+-magadiite and used in a study of mixed-metal bioactive glasses.
Specifications
Specifications
| CAS | 78-10-4 |
| Density | 0.933 g/mL (at 20°C) |
| Boiling Point | 168°C |
| Molecular Formula | C8H20O4Si |
| Refractive Index | n20/D 1.382 (literature) |
| Linear Formula | Si(OC2H5)4 |
| Quantity | 4 L |
| MDL Number | MFCD00009062 |
| Synonym | Orthosilicic acid tetraethyl ester; Silicon tetraethoxide; TEOS; Tetraethoxysilane; Tetraethoxysilicon(IV); Tetraethyl silicate; Orthosilicic acid tetraethyl ester; TEOS; Tetraethoxysilane |
| Molecular Weight (g/mol) | 208.33 |
| Show More |