Learn More
Tetraethyl orthocarbonate, 97+%
CAS: 78-09-1 | C9H20O4 | 192.26 g/mol
$287.74 - $962.10
Chemical Identifiers
| CAS | 78-09-1 |
|---|---|
| Molecular Formula | C9H20O4 |
| Molecular Weight (g/mol) | 192.26 |
| MDL Number | MFCD00009221 |
| InChI Key | CWLNAJYDRSIKJS-UHFFFAOYSA-N |
| Synonym | tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane |
| PubChem CID | 66213 |
| IUPAC Name | triethoxymethoxyethane |
| SMILES | CCOC(OCC)(OCC)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1783514
|
Thermo Scientific Chemicals
A1783514 |
25 g |
Each for $287.74
|
|
|||||
|
AAA1783522
|
Thermo Scientific Chemicals
A1783522 |
100 g |
Each for $962.10
|
|
|||||
Description
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
Solubility
Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol.
Notes
Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert atmosphere and refrigerated.
Chemical Identifiers
| 78-09-1 | |
| 192.26 | |
| CWLNAJYDRSIKJS-UHFFFAOYSA-N | |
| 66213 | |
| CCOC(OCC)(OCC)OCC |
| C9H20O4 | |
| MFCD00009221 | |
| tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane | |
| triethoxymethoxyethane |
Specifications
| 78-09-1 | |
| 158°C to 161°C | |
| C9H20O4 | |
| MFCD00009221 | |
| UN3272 | |
| Moisture sensitive | |
| Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol. | |
| CCOC(OCC)(OCC)OCC | |
| 192.26 | |
| 192.26 | |
| Tetraethyl orthocarbonate |
| 0.917 | |
| 52°C (125°F) | |
| 1.393 | |
| 25 g | |
| 1747401 | |
| tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane | |
| CWLNAJYDRSIKJS-UHFFFAOYSA-N | |
| triethoxymethoxyethane | |
| 66213 | |
| ≥98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 201-082-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only